Methyl-3-(trifluoromethyl)benzoate ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
Methyl 3-(trifluoromethyl)benzoate
M1718-25G
M1718-5G
TCM1718-25GEA
43.9
USD
TCM1718-25G
TCM1718-5G
Methyl-3-(trifluoromethyl)benzoate ≥98.0% (by GC)
Methyl-3-(trifluoromethyl)benzoate
CAS Number: 2557-13-3
Molecular Formula: C9H7F3O2
Molecular Weight: 204.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 198
Flash Point (°C): 40
Specific Gravity (20/20): 1.30
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Methyl alpha,alpha,alpha-Trifluoro-m-toluate, 3-(Trifluoromethyl)benzoic Acid Methyl Ester
Formula:
C₉H₇F₃O₂ MW: 204.15 g/mol Boiling Pt: 198 °C Density: 1.30 (20 °C) Flash Pt: 40 °C |
MDL Number:
MFCD00043543 CAS Number: 2557-13-3 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...