4-Nitro-m-toluidine ≥97.0% (by GC)
Supplier: TCI America
Synonyms:
3-Methyl-4-nitroaniline, 4-Nitro-3-methylaniline, 5-Amino-2-nitrotoluene
M1677-25G
M1677-5G
TCM1677-25GEA
194.6
USD
TCM1677-25G
TCM1677-5G
4-Nitro-m-toluidine ≥97.0% (by GC)
4-Nitro-m-toluidine
CAS Number: 611-05-2
Molecular Formula: C7H8N2O2
Molecular Weight: 152.15
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 134
Product Categories: Materials Chemistry, Organic Nonlinear Optical (NLO) Materials
Other Product Names / Synonyms: 5-Amino-2-nitrotoluene, 4-Nitro-m-toluidine
Formula:
C₇H₈N₂O₂ MW: 152.15 g/mol Melting Pt: 134 °C |
MDL Number:
MFCD00091833 CAS Number: 611-05-2 UN: 2660 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...