(±)-5-Methyl-2-hexanol ≥98.0% (by GC)

Supplier: TCI America

M1666-25G M1666-5G
TCM1666-25GEA 82 USD
TCM1666-25G TCM1666-5G
(±)-5-Methyl-2-hexanol ≥98.0% (by GC)
(±)-5-Methyl-2-hexanol

CAS Number: 627-59-8
Molecular Formula: C7H16O
Molecular Weight: 116.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 150
Flash Point (°C): 46
Specific Gravity (20/20): 0.81

Formula: C₇H₁₆O
MW: 116.2 g/mol
Boiling Pt: 150 °C
Density: 0.81 (20 °C)
Flash Pt: 46 °C
MDL Number: MFCD00004558
CAS Number: 627-59-8
UN: 1987
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3901bbc5a59f41afe1fc1a00a0b35fd2ac4d6585&vid=3901bbc5a59f41afe1fc1a00a0b35fd2ac4d6585&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR