Milrinone ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: 6-Methyl-5-(4-pyridyl)-2-pyridone-3-carbonitrile, 2-Methyl-6-oxo-1,6-dihydro-[3,4'-bipyridine]-5-carbonitrile, 3-Cyano-6-methyl-5-(4-pyridyl)-2-pyridone

M1663-100MG M1663-1G
TCM1663-100MGEA 102 USD
TCM1663-100MG TCM1663-1G
Milrinone ≥98.0% (by total nitrogen basis)
Milrinone

CAS Number: 78415-72-2
MDL Number: MFCD00133539
Molecular Formula: C12H9N3O
Molecular Weight: 211.22
Purity/Analysis Method: >98.0% (N)
Form: Crystal


Product Categories: Bioscience, Inhibitors (for Pharmacological Research)
Other Product Names / Synonyms: 6-Methyl-5-(4-pyridyl)-2-pyridone-3-carbonitrile, 3-Cyano-6-methyl-5-(4-pyridyl)-2-pyridone

Formula: C₁₂H₉N₃O
MW: 211.22 g/mol
MDL Number: MFCD00133539
CAS Number: 78415-72-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c797f2007ee9d8a00b4a9e9a92726e794d93c91c&vid=c797f2007ee9d8a00b4a9e9a92726e794d93c91c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR