(3-Mercaptopropyl)triethoxysilane ≥96.0% (by GC)
Supplier: TCI America
Synonyms:
3-(Triethoxysilyl)propanethiol, 3-Mercaptopropyltriethoxysilane
M1505-25G
M1505-500G
TCM1505-25GEA
35.6
USD
TCM1505-25G
TCM1505-500G
(3-Mercaptopropyl)triethoxysilane ≥96.0% (by GC)
(3-Mercaptopropyl)triethoxysilane
CAS Number: 14814-09-6
MDL Number: MFCD00053924
Molecular Formula: C9H22O3SSi
Molecular Weight: 238.42
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 115
Flash Point (°C): 88
Specific Gravity (20/20): 0.99
Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: 3-(Triethoxysilyl)propanethiol
Formula:
HSH₂CCH₂CH₂Si(OCH₂CH₃)₃ MW: 238.42 g/mol Boiling Pt: 115 °C Density: 0.99 (20 °C) Flash Pt: 88 °C |
MDL Number:
MFCD00053924 CAS Number: 14814-09-6 |
Specification Test Results
Purity/Analysis Method | >96.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...