2-Methoxy-1-butanol ≥98.0% (by GC)

Supplier: TCI America

M1497-25G M1497-5G
TCM1497-25GEA 558 USD
TCM1497-25G TCM1497-5G
2-Methoxy-1-butanol ≥98.0% (by GC)
2-Methoxy-1-butanol

CAS Number: 15467-25-1
Molecular Formula: C5H12O2
Molecular Weight: 104.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 146
Specific Gravity (20/20): 0.93

Formula: C₅H₁₂O₂
MW: 104.15 g/mol
Boiling Pt: 146 °C
Density: 0.93 (20 °C)
CAS Number: 15467-25-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=839966f3bbfca1d50b679a43c8606fdb672ecb8a&vid=839966f3bbfca1d50b679a43c8606fdb672ecb8a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR