3-Methoxy-o-toluidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M1493-25G M1493-5G
TCM1493-25GEA 331 USD
TCM1493-25G TCM1493-5G
3-Methoxy-o-toluidine ≥98.0% (by GC, titration analysis)
3-Methoxy-o-toluidine

CAS Number: 19500-02-8
MDL Number: MFCD06412568
Molecular Formula: C8H11NO
Molecular Weight: 137.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 1.08
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 1-Amino-3-methoxy-2-methylbenzene, 3-Methoxy-o-toluidine

Formula: C₈H₁₁NO
Density: 1.08 (20 °C)
MDL Number: MFCD06412568
CAS Number: 19500-02-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05a1877fab8f7204d47107a79f782c89dc04c8bc&vid=05a1877fab8f7204d47107a79f782c89dc04c8bc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR