3-Methoxy-1-propanol ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
Trimethylene glycol monomethyl ether
M1475-250G
M1475-25G
TCM1475-250GEA
126
USD
TCM1475-250G
TCM1475-25G
3-Methoxy-1-propanol ≥98.0% (by GC)
3-Methoxy-1-propanol
CAS Number: 1589-49-7
MDL Number: MFCD00039550
Molecular Formula: C4H10O2
Molecular Weight: 90.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 153
Flash Point (°C): 38
Specific Gravity (20/20): 0.94
Other Product Names / Synonyms: 1,3-Propanediol Monomethyl Ether
Formula:
C₄H₁₀O₂ MW: 90.12 g/mol Boiling Pt: 153 °C Density: 0.94 (20 °C) Flash Pt: 38 °C |
MDL Number:
MFCD00039550 CAS Number: 1589-49-7 UN: 1987 ADR: 3,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...