4-Methoxy-o-toluidine ≥99.0%

Supplier: TCI America

Synonyms: 4-Methoxy-2-methylaniline, m-Cresidine, 2-Methyl-p-anisidine, 1-Amino-4-methoxy-2-methylbenzene, 2-Amino-5-methoxytoluene, 4-Amino-3-methylanisole

M1474-25G M1474-500G
TCM1474-25GEA 101 USD
TCM1474-25G TCM1474-500G
4-Methoxy-o-toluidine ≥99.0%
4-Methoxy-o-toluidine

CAS Number: 102-50-1
MDL Number: MFCD00007735
Molecular Formula: C8H11NO
Molecular Weight: 137.18
Purity/Analysis Method: >99.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 249
Melting point (°C): 13
Flash Point (°C): 110
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 1-Amino-4-methoxy-2-methylbenzene, m-Cresidine, 4-Methoxy-o-toluidine

Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 248
Melting Pt: 13 °C
Flash Pt: 110 °C
MDL Number: MFCD00007735
CAS Number: 102-50-1
UN: 2810
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d368cf785b18fcb7bba2079074ef74d34d2ff1c2&vid=d368cf785b18fcb7bba2079074ef74d34d2ff1c2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR