Methyl-3-mercaptopropionate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 3-Mercaptopropionic acid methyl ester, Methyl 3-mercaptopropanoate

M1434-25G M1434-500G
TCM1434-25GEA 75 USD
TCM1434-25G TCM1434-500G
Methyl-3-mercaptopropionate ≥98.0% (by GC)
Methyl-3-mercaptopropionate

CAS Number: 2935-90-2
MDL Number: MFCD00004895
Molecular Formula: C4H8O2S
Molecular Weight: 120.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 64
Specific Gravity (20/20): 1.11


Other Product Names / Synonyms: 3-Mercaptopropionic Acid Methyl Ester

Formula: C₄H₈O₂S
MW: 120.17 g/mol
Density: 1.11 (20 °C)
Flash Pt: 64 °C
MDL Number: MFCD00004895
CAS Number: 2935-90-2
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8aa77197e413a2d8504cf31235bf0fe170dc8968&vid=8aa77197e413a2d8504cf31235bf0fe170dc8968&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR