1-Methylpyridinium chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

M1397-25G M1397-5G
TCM1397-25GEA 387 USD
TCM1397-25G TCM1397-5G
1-Methylpyridinium chloride ≥98.0% (by titrimetric analysis)
1-Methylpyridinium chloride

CAS Number: 7680-73-1
MDL Number: MFCD00137532
Molecular Formula: C6H8ClN
Molecular Weight: 129.59
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 144

Formula: C₆H₈ClN
Melting Pt: 144 °C
MDL Number: MFCD00137532
CAS Number: 7680-73-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e0907f268b49fe1bcba25f1bb70f4ec06e8e97f&vid=4e0907f268b49fe1bcba25f1bb70f4ec06e8e97f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR