3-Methoxybenzonitrile ≥95.0% (by GC)
Supplier: TCI America
CAS Number: 1527-89-5
MDL Number: MFCD00001801
Molecular Formula: C8H7NO
Molecular Weight: 133.15
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 140
Flash Point (°C): 105
Freezing point (°C): 22
Other Product Names / Synonyms: 3-Cyanoanisole
Formula:
C₈H₇NO Boiling Pt: 140 °C Flash Pt: 105 °C |
MDL Number:
MFCD00001801 CAS Number: 1527-89-5 UN: 3439 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...