3-Methylbenzylamine ≥97.0% (by GC, titration analysis)
Supplier: TCI America
CAS Number: 100-81-2
MDL Number: MFCD00008118
Molecular Formula: C8H11N
Molecular Weight: 121.18
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 206
Flash Point (°C): 81
Specific Gravity (20/20): 0.97
Other Product Names / Synonyms: alpha-Amino-m-xylene
Formula:
C₈H₁₁N Boiling Pt: 206 °C Density: 0.97 (20 °C) Flash Pt: 81 °C |
MDL Number:
MFCD00008118 CAS Number: 100-81-2 UN: 2735 ADR: 8,III |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...