Nitro-tert-butane ≥98.0% (by GC)

Supplier: TCI America

M1241-25G M1241-5G
TCM1241-25GEA 205 USD
TCM1241-25G TCM1241-5G
Nitro-tert-butane ≥98.0% (by GC)
Nitro-tert-butane

CAS Number: 594-70-7
MDL Number: MFCD00007393
Molecular Formula: C4H9NO2
Molecular Weight: 103.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Flash Point (°C): 19
Freezing point (°C): 24

Formula: C₄H₉NO₂
Flash Pt: 19 °C
MDL Number: MFCD00007393
CAS Number: 594-70-7
UN: 1325
ADR: 4.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09c517a139791ab8145128591a9061d91bab7260&vid=76876306fe6c79b0ea94040964c9f8d941f7ae94&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR