α,α,α-Trifluoro-o-xylene ≥98.0% (by GC)
Supplier: TCI America
CAS Number: 13630-19-8
MDL Number: MFCD00153204
Molecular Formula: C8H7F3
Molecular Weight: 160.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 126
Specific Gravity (20/20): 1.17
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-(Trifluoromethyl)toluene, alpha,alpha,alpha-Trifluoro-o-xylene
Formula:
C₈H₇F₃ Boiling Pt: 126 °C Density: 1.17 (20 °C) |
MDL Number:
MFCD00153204 CAS Number: 13630-19-8 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...