(R)-(-)-2-(Methoxymethyl)pyrrolidine ≥99.0% (by GC)
Supplier: TCI America
Synonyms:
(R)-2-(Methoxymethyl)pyrrolidine
M1169-1G
M1169-5G
TCM1169-1GEA
105.7
USD
TCM1169-1G
TCM1169-5G
(R)-(-)-2-(Methoxymethyl)pyrrolidine ≥99.0% (by GC)
(R)-(-)-2-(Methoxymethyl)pyrrolidine
CAS Number: 84025-81-0
MDL Number: MFCD00066219
Molecular Formula: C6H13NO
Molecular Weight: 115.18
Purity/Analysis Method:
Formula:
C₆H₁₃NO MW: 115.18 g/mol Density: 0.93 (20 °C) Flash Pt: 45 °C |
MDL Number:
MFCD00066219 CAS Number: 84025-81-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...