(S)-(+)-2-(Methoxymethyl)pyrrolidine ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (S)-2-(Methoxymethyl)pyrrolidine

M1161-1G M1161-5G
TCM1161-1GEA 66 USD
TCM1161-1G TCM1161-5G
(S)-(+)-2-(Methoxymethyl)pyrrolidine ≥98.0% (by GC)
(S)-(+)-2-(Methoxymethyl)pyrrolidine

CAS Number: 63126-47-6 MDL Number: MFCD00010408 Molecular Formula: C6H13NO Molecular Weight: 115.18 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 68 Flash Point (°C): 45 Specific Gravity (20/20): 0.94 Specific rotation [a]20/D: 38 deg (C=2, 1mol/L HCl)

Formula: C₆H₁₃NO
MW: 115.18 g/mol
Boiling Pt: 68 °C
Density: 0.94 (20 °C)
Flash Pt: 45 °C
MDL Number: MFCD00010408
CAS Number: 63126-47-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a30b1fd30bcd6b23c0bfd8ffc982b5541323b10&vid=6a30b1fd30bcd6b23c0bfd8ffc982b5541323b10&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR