2-Methylresorcinol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2,6-Dihydroxytoluene, 2-Methylbenzene-1,3-diol

M1109-25G M1109-500G
TCM1109-25GEA 69 USD
TCM1109-25G TCM1109-500G
2-Methylresorcinol ≥98.0% (by GC)
2-Methylresorcinol

CAS Number: 608-25-3
MDL Number: MFCD00002271
Molecular Formula: C7H8O2
Molecular Weight: 124.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 122
Flash Point (°C): 135


Other Product Names / Synonyms: 2,6-Dihydroxytoluene

Formula: C₇H₈O₂
MW: 124.14 g/mol
Melting Pt: 122 °C
Flash Pt: 135 °C
MDL Number: MFCD00002271
CAS Number: 608-25-3
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=782b0e21a8014b6a95d4d2526f4a8d0db7c9a9fa&vid=782b0e21a8014b6a95d4d2526f4a8d0db7c9a9fa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR