1-Methyl-1H-pyrazole ≥98.0% (by GC)

Supplier: TCI America

M0969-25G M0969-5G
TCM0969-25GEA 84 USD
TCM0969-25G TCM0969-5G
1-Methyl-1H-pyrazole ≥98.0% (by GC)
1-Methyl-1H-pyrazole

CAS Number: 930-36-9
MDL Number: MFCD00144943
Molecular Formula: C4H6N2
Molecular Weight: 82.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 127
Flash Point (°C): 36
Specific Gravity (20/20): 0.99

Formula: C₄H₆N₂
Boiling Pt: 127 °C
Density: 0.99 (20 °C)
Flash Pt: 36 °C
MDL Number: MFCD00144943
CAS Number: 930-36-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d74a412cd1bebf2c82676d6f9ca18474eb08aa3d&vid=d74a412cd1bebf2c82676d6f9ca18474eb08aa3d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR