(R)-(-)-3-Chloro-1,2-propanediol ≥96.0% (by GC)

Supplier: TCI America

Synonyms: (R)-(-)-3-Chloro-1,2-dihydroxy propane, (2R)-(-)-3-Chloropropane-1,2-diol

M0967-10G M0967-25G
TCM0967-10GEA 74 USD
TCM0967-10G TCM0967-25G
(R)-(-)-3-Chloro-1,2-propanediol ≥96.0% (by GC)
(R)-(-)-3-Chloro-1,2-propanediol

CAS Number: 57090-45-6
MDL Number: MFCD00135169
Molecular Formula: C3H7ClO2
Molecular Weight: 110.54
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 95
Flash Point (°C): 126
Specific Gravity (20/20): 1.32
Specific rotation [a]20/D: -9 deg (C=2, H2O)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(-)-alpha-Chlorohydrin, Glycerol (R)-(-)-alpha-Monochlorohydrin, (R)-(-)-alpha-Monochlorohydrin

Formula: ClCH₂CHOHCH₂OH
MW: 110.54 g/mol
Boiling Pt: 95 °C
Density: 1.32 (20 °C)
Flash Pt: 126 °C
MDL Number: MFCD00135169
CAS Number: 57090-45-6
UN: 2689
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c93f34937887b5d732f683f9754e102a9ed6651&vid=0c93f34937887b5d732f683f9754e102a9ed6651&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR