(±)-2-Methylcaproic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-MethylhexanoicAcid, 2-Methylhexanoic acid, (±)-2-Methylhexanoic acid

M0947-25ML M0947-5ML
TCM0947-25MLEA 120 USD
TCM0947-25ML TCM0947-5ML
(±)-2-Methylcaproic acid ≥98.0% (by GC, titration analysis)
(±)-2-Methylcaproic acid

CAS Number: 4536-23-6
MDL Number: MFCD00002674
Molecular Formula: C7H14O2
Molecular Weight: 130.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 72
Flash Point (°C): 105
Specific Gravity (20/20): 0.92

Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 72 °C
Density: 0.92 (20 °C)
Flash Pt: 105 °C
MDL Number: MFCD00002674
CAS Number: 4536-23-6
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e28b74ad482d50b8fa0af149f37628694a97498&vid=8e28b74ad482d50b8fa0af149f37628694a97498&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR