2-Methylbenzylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M0942-25ML M0942-5ML
TCM0942-25MLEA 91 USD
TCM0942-25ML TCM0942-5ML
2-Methylbenzylamine ≥98.0% (by GC, titration analysis)
2-Methylbenzylamine

CAS Number: 89-93-0
MDL Number: MFCD00008112
Molecular Formula: C8H11N
Molecular Weight: 121.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 82
Flash Point (°C): 84
Specific Gravity (20/20): 0.98


Other Product Names / Synonyms: alpha-Amino-o-xylene

Formula: C₈H₁₁N
Boiling Pt: 82 °C
Density: 0.98 (20 °C)
Flash Pt: 84 °C
MDL Number: MFCD00008112
CAS Number: 89-93-0
UN: 2735
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c60a5b2f601f418d379e176e027b3ed196a5644a&vid=c60a5b2f601f418d379e176e027b3ed196a5644a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR