Methyl dichlorophosphate ≥95.0%
Supplier: TCI America
Synonyms:
Methyl phosphorodichloridate, Methyl dichlorophosphate
M0904-100G
M0904-25G
TCM0904-100GEA
599
USD
TCM0904-100G
TCM0904-25G
TCM0904-5G
Methyl dichlorophosphate ≥95.0%
Methyl dichlorophosphate
CAS Number: 677-24-7
MDL Number: MFCD00002068
Molecular Formula: CH3Cl2O2P
Molecular Weight: 148.91
Purity/Analysis Method: >95.0% (T)
Form: Clear Liquid
Boiling point (°C): 63
Flash Point (°C): 110
Specific Gravity (20/20): 1.50
Product Categories: Synthetic Organic Chemistry, Phosphorylation, Nucleic Acid Synthesis
Other Product Names / Synonyms: Methyl Dichlorophosphate
Formula:
CH₃Cl₂O₂P MW: 148.91 g/mol Boiling Pt: 63 °C Density: 1.50 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00002068 CAS Number: 677-24-7 UN: 3265 ADR: 8 III |
|
Specification Test Results
Purity/Analysis Method | >95.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...