Ethyl (methylthio)acetate ≥98.0%
Supplier: TCI America
Synonyms:
(Methylthio)acetic ethyl ester, Ethyl 2-(methylthio)acetate
M0897-25G
M0897-5G
TCM0897-25GEA
52
USD
TCM0897-25G
TCM0897-5G
Ethyl (methylthio)acetate ≥98.0%
Ethyl (methylthio)acetate
CAS Number: 4455-13-4
MDL Number: MFCD00009182
Molecular Formula: C5H10O2S
Molecular Weight: 134.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 176
Flash Point (°C): 59
Specific Gravity (20/20): 1.06
Product Categories: Synthetic Organic Chemistry, Sulfides
Other Product Names / Synonyms: (Methylthio)acetic Acid Ethyl Ester
Formula:
C₅H₁₀O₂S Boiling Pt: 176 °C Density: 1.06 (20 °C) Flash Pt: 59 °C |
MDL Number:
MFCD00009182 CAS Number: 4455-13-4 UN: 3272 ADR: 3,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...