O-Methylhydroxylamine hydrochloride ∼40% ∼5.4 mol/L in water

Supplier: TCI America

Synonyms: Methylhydroxylamine hydrochloride, O-Methylhydroxylammonium chloride, Methoxyamine hydrochloride, Methoxylamine hydrochloride

M0886-25ML M0886-500ML
TCM0886-25MLEA 33 USD
TCM0886-25ML TCM0886-500ML
O-Methylhydroxylamine hydrochloride ∼40% ∼5.4 mol/L in water
O-Methylhydroxylamine hydrochloride

CAS Number: 593-56-6
MDL Number: MFCD00012951
Molecular Formula: CH5NO
Molecular Weight: 83.52
Form: Clear Liquid
Color: Colorless


Other Product Names / Synonyms: Methoxyamine Hydrochloride, Methoxylamine Hydrochloride

Formula: CH₆ClNO
MW: 83.515 g/mol
MDL Number: MFCD00012951
CAS Number: 593-56-6
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0cd506adfb72dcdf1523f1f9e591801ae08b719c&vid=0a70e6a58bf3a9d44b3655e2255baeb73e3e4314&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR