2-Methyl-2-oxazoline ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-Methyl-4,5-dihydrooxazole

M0857-25G M0857-500G
TCM0857-25GEA 43 USD
TCM0857-25G TCM0857-500G
2-Methyl-2-oxazoline ≥98.0% (by GC)
2-Methyl-2-oxazoline

CAS Number: 1120-64-5
MDL Number: MFCD00005298
Molecular Formula: C4H7NO
Molecular Weight: 85.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 110
Flash Point (°C): 20
Specific Gravity (20/20): 1.01

Formula: C₄H₇NO
MW: 85.11 g/mol
Boiling Pt: 110 °C
Density: 1.01 (20 °C)
Flash Pt: 20 °C
MDL Number: MFCD00005298
CAS Number: 1120-64-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=960a844cdadc276e2c45100ec06e27fbe1d95293&vid=960a844cdadc276e2c45100ec06e27fbe1d95293&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR