3-Methoxybenzyl chloride ≥96.0%
Supplier: TCI America
Synonyms:
1-(Chloromethyl)-3-methoxybenzene, 3-(Chloromethyl)anisole
M0820-25G
M0820-5G
TCM0820-25GEA
87
USD
TCM0820-25G
TCM0820-5G
3-Methoxybenzyl chloride ≥96.0%
3-Methoxybenzyl chloride
CAS Number: 824-98-6
MDL Number: MFCD00000907
Molecular Formula: C8H9ClO
Molecular Weight: 156.61
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 125
Flash Point (°C): 101
Specific Gravity (20/20): 1.16
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 3-(Chloromethyl)anisole
Formula:
C₈H₉ClO Boiling Pt: 125 °C Density: 1.16 (20 °C) Flash Pt: 101 °C |
MDL Number:
MFCD00000907 CAS Number: 824-98-6 UN: 1760 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method | >96.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...