ω-Bromo-3-methoxyacetophenone ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2-Bromo-3'-methoxyacetophenone, Bromomethyl m-methoxyphenyl ketone, 3-Methoxyphenacyl bromide, 2-Bromo-1-(3-methoxyphenyl)ethanone, 2-Bromo-1-(3-methoxyphenyl)ethan-1-one, α-Bromo-3-methoxyacetophenone
CAS Number: 5000-65-7
MDL Number: MFCD00000199
Molecular Formula: C9H9BrO2
Molecular Weight: 229.07
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 63
Product Categories: Analytical Science, UV Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: 3-(Bromoacetyl)anisole, 2-Bromo-3'-methoxyacetophenone
Formula:
C₉H₉BrO₂ MW: 229.07 g/mol Melting Pt: 63 °C |
MDL Number:
MFCD00000199 CAS Number: 5000-65-7 UN: 3261 ADR: 8,II |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...