2-Methoxy-3-methylpyrazine ≥98.0% (by GC)

Supplier: TCI America

M0810-25G M0810-5G
TCM0810-25GEA 122.4 USD
TCM0810-25G TCM0810-5G
2-Methoxy-3-methylpyrazine ≥98.0% (by GC)
2-Methoxy-3-methylpyrazine

CAS Number: 2847-30-5
MDL Number: MFCD00006127
Molecular Formula: C6H8N2O
Molecular Weight: 124.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 87
Flash Point (°C): 56
Specific Gravity (20/20): 1.08

Formula: C₆H₈N₂O
Boiling Pt: 87 °C
Density: 1.08 (20 °C)
Flash Pt: 56 °C
MDL Number: MFCD00006127
CAS Number: 2847-30-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d284ce729f606d970245596a21a9f2ff2a074d22&vid=d284ce729f606d970245596a21a9f2ff2a074d22&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR