2-Methyl-2-pentenoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
M0784-25G
M0784-5G
TCM0784-25GEA
49
USD
TCM0784-25G
TCM0784-5G
2-Methyl-2-pentenoic acid ≥98.0% (by GC, titration analysis)
2-Methyl-2-pentenoic acid
CAS Number: 3142-72-1
MDL Number: MFCD00002655
Molecular Formula: C6H10O2
Molecular Weight: 114.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Flash Point (°C): 107
Freezing point (°C): 23
Formula:
C₆H₁₀O₂ Flash Pt: 107 °C |
MDL Number:
MFCD00002655 CAS Number: 3142-72-1 UN: 3265 ADR: 8,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...