2-Methylpyrazine ≥98.0% (by GC)

Supplier: TCI America

M0736-250ML M0736-25ML
TCM0736-250MLEA 127.6 USD
TCM0736-250ML TCM0736-25ML
2-Methylpyrazine ≥98.0% (by GC)
2-Methylpyrazine

CAS Number: 109-08-0
MDL Number: MFCD00006142
Molecular Formula: C5H6N2
Molecular Weight: 94.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 135
Melting point (°C): -29
Flash Point (°C): 50
Specific Gravity (20/20): 1.03

Formula: C₅H₆N₂
MW: 94.12 g/mol
Boiling Pt: 135 °C
Melting Pt: –29 °C
Density: 1.03 (20 °C)
Flash Pt: 50 °C
MDL Number: MFCD00006142
CAS Number: 109-08-0
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=310d3ae6eb3889ee2779637bc9f36a98f60e62b4&vid=310d3ae6eb3889ee2779637bc9f36a98f60e62b4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR