2-Methyl-2-butene ≥90.0% (by GC)

Supplier: TCI America

Synonyms: Trimethylethylene, Isoamylene, Isopentene

M0708-25ML M0708-500ML M0708-100ML
TCM0708-25MLEA 25 USD
TCM0708-25ML TCM0708-500ML TCM0708-100ML
2-Methyl-2-butene ≥90.0% (by GC)
2-Methyl-2-butene

CAS Number: 513-35-9
MDL Number: MFCD00009276
Molecular Formula: C5H10
Molecular Weight: 70.14
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 38
Melting point (°C): -134
Flash Point (°C): -45
Specific Gravity (20/20): 0.66


Other Product Names / Synonyms: Amylene, Pentene

Formula: C₅H₁₀
MW: 70.13 g/mol
Boiling Pt: 38 °C
Melting Pt: -134 °C
Density: 0.66 (20 °C)
Flash Pt: -45 °C
MDL Number: MFCD00009276
CAS Number: 513-35-9
UN: 2460
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15f70fcdc73bdd846ca166c74801b57acabf1bde&vid=48a4d1e0167519114dd3207114e5524cfca7d751&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR