Methoxytrimethylsilane ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Trimethyl(methoxy)silane

M0688-250ML M0688-25ML
TCM0688-250MLEA 174 USD
TCM0688-250ML TCM0688-25ML
Methoxytrimethylsilane ≥98.0% (by GC)
Methoxytrimethylsilane

CAS Number: 1825-61-2
MDL Number: MFCD00008344
Molecular Formula: C4H12OSi
Molecular Weight: 104.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 57
Flash Point (°C): -30
Specific Gravity (20/20): 0.76

Formula: C₄H₁₂OSi
MW: 104.22 g/mol
Boiling Pt: 57 °C
Density: 0.76 (20 °C)
Flash Pt: -30 °C
MDL Number: MFCD00008344
CAS Number: 1825-61-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c38024f1fa4a16de76e2131e01a9da6e69176b8&vid=4c38024f1fa4a16de76e2131e01a9da6e69176b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR