N-Methyl-N-(trimethylsilyl)trifluoroacetamide ≥95.0% (by GC)

Supplier: TCI America

M0672-25ML M0672-5ML
TCM0672-25MLEA 177.7 USD
TCM0672-25ML TCM0672-5ML
N-Methyl-N-(trimethylsilyl)trifluoroacetamide ≥95.0% (by GC)
N-Methyl-N-(trimethylsilyl)trifluoroacetamide

[Trimethylsilylating Agent]
CAS Number: 24589-78-4
MDL Number: MFCD00000411
Molecular Formula: C6H12F3NOSi
Molecular Weight: 199.25
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 132
Flash Point (°C): 26
Specific Gravity (20/20): 1.08


Product Categories: Analytical Science, Silylation (GC Derivatizing Reagents)
Other Product Names / Synonyms: MSTFA

Formula: C₆H₁₂F₃NOSi
Boiling Pt: 132 °C
Density: 1.08 (20 °C)
Flash Pt: 26 °C
MDL Number: MFCD00000411
CAS Number: 24589-78-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a676142f1b6f92f6205ace284af31c85a2c0635&vid=fca06cc9ef135fe86a98d711d7401fdb5eee4448&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR