3-Methyl-2-pentanone ≥98.0% (by GC)
Supplier: TCI America
M0388-25ML
M0388-5ML
TCM0388-25MLEA
136
USD
TCM0388-25ML
TCM0388-5ML
3-Methyl-2-pentanone ≥98.0% (by GC)
3-Methyl-2-pentanone
CAS Number: 565-61-7
MDL Number: MFCD00009336
Molecular Formula: C6H12O
Molecular Weight: 100.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 116
Flash Point (°C): 12
Specific Gravity (20/20): 0.81
Other Product Names / Synonyms: sec-Butyl Methyl Ketone
Formula:
C₆H₁₂O MW: 100.16 g/mol Boiling Pt: 116 °C Density: 0.81 (20 °C) Flash Pt: 12 °C |
MDL Number:
MFCD00009336 CAS Number: 565-61-7 UN: 1224 ADR: 3,II |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...