2-Methyl-2-thiazoline ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
2-Methyl-4,5-dihydrothiazole
M0285-100ML
M0285-25ML
TCM0285-100MLEA
433
USD
TCM0285-100ML
TCM0285-25ML
2-Methyl-2-thiazoline ≥98.0% (by GC)
2-Methyl-2-thiazoline
CAS Number: 2346-00-1
MDL Number: MFCD00005314
Molecular Formula: C4H7NS
Molecular Weight: 101.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 145
Flash Point (°C): 37
Specific Gravity (20/20): 1.08
Storage Temperature: 0-10°C
Formula:
C₄H₇NS MW: 101.17 g/mol Boiling Pt: 145 °C Density: 1.08 (20 °C) Flash Pt: 37 °C |
MDL Number:
MFCD00005314 CAS Number: 2346-00-1 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...