4-Methyl-2-pentenoic acid ≥97.0% (by GC, titration analysis) stabilized

Supplier: TCI America

M0268-25ML M0268-5ML
TCM0268-25MLEA 339 USD
TCM0268-25ML TCM0268-5ML
4-Methyl-2-pentenoic acid ≥97.0% (by GC, titration analysis) stabilized
4-Methyl-2-pentenoic acid

(stabilized with HQ)
CAS Number: 10321-71-8
MDL Number: MFCD00043804
Molecular Formula: C6H10O2
Molecular Weight: 114.14
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 212
Flash Point (°C): 110
Specific Gravity (20/20): 0.96
Storage Temperature: 0-10°C

Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 212 °C
Density: 0.96 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00043804
CAS Number: 10321-71-8
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be5beac5890ecedb80db9ad3777b7507f626745d&vid=be5beac5890ecedb80db9ad3777b7507f626745d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR