Methyl red - Methylene blue in ethanol

Supplier: TCI America

Synonyms: Methylene Blue - Methyl Red Solution

M0161-100ML M0161-500ML
TCM0161-100MLEA 49 USD
TCM0161-100ML TCM0161-500ML
Methyl red - Methylene blue in ethanol
Methyl red - Methylene blue

[mixed Indicator]
MDL Number: MFCD00079686
Form: Liquid
Color: Deep Green
Flash Point (°C): 13


Other Product Names / Synonyms: Methylene Blue - Methyl Red Solution

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR