Lornoxicam ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
CAS Number: 70374-39-9
MDL Number: MFCD00866163
Molecular Formula: C13H10ClN3O4S2
Molecular Weight: 371.81
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 230
Storage Temperature: >-20°C
Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Inhibitors (for Pharmacological Research), Anti-rheumatoid Arthritis Ingredients, Anti-inflammatory Ingredients, Anti-inflammatory Ingredients
Other Product Names / Synonyms: 6-Chloro-4-hydroxy-2-methyl-N-(2-pyridyl)-2H-thieno[2,3-e][1,2]thiazine-3-carboxamide 1,1-Dioxide
Formula:
C₁₃H₁₀ClN₃O₄S₂ Melting Pt: 230 °C |
MDL Number:
MFCD00866163 CAS Number: 70374-39-9 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,N) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...