Lithium aluminum hydride ≥98.0% (by titrimetric analysis), powder
Supplier: TCI America
Synonyms:
Aluminium lithium hydride, Lithium tetrahydridoaluminate
L0203-100G
L0203-25G
TCL0203-100GEA
442.29
USD
TCL0203-100G
TCL0203-25G
Lithium aluminum hydride ≥98.0% (by titrimetric analysis), powder
Lithium aluminum hydride
CAS Number: 16853-85-3
MDL Number: MFCD00011075
Molecular Formula: H4AlLi
Molecular Weight: 37.95
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 125
Product Categories: Synthetic Organic Chemistry, Reduction, Materials Chemistry, Fuel Cell Research Reagents
Other Product Names / Synonyms: LAH
Formula:
H₄AlLi MW: 37.95 g/mol Melting Pt: 125 °C |
MDL Number:
MFCD00011075 CAS Number: 16853-85-3 UN: 1410 ADR: 4.3,I |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...