3,4-Lutidine ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 3,4-Dimethylpyridine

L0066-25ML L0066-500ML
TCL0066-25MLEA 32 USD
TCL0066-25ML TCL0066-500ML
3,4-Lutidine ≥98.0% (by GC)
3,4-Lutidine

CAS Number: 583-58-4
MDL Number: MFCD00006403
Molecular Formula: C7H9N
Molecular Weight: 107.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 179
Melting point (°C): -6
Flash Point (°C): 61
Specific Gravity (20/20): 0.96


Product Categories: Synthetic Organic Chemistry, Ligands, Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: 3,4-Dimethylpyridine

Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 179 °C
Melting Pt: -6 °C
Density: 0.96 (20 °C)
Flash Pt: 61 °C
MDL Number: MFCD00006403
CAS Number: 583-58-4
UN: 2929
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3aae8974df7f7b3e5e618265df81c5a623742e50&vid=3aae8974df7f7b3e5e618265df81c5a623742e50&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR