Ketoconazole ≥98.0%
Supplier: TCI America
Synonyms:
(+/-)-cis-1-Acetyl-4-[4-[[2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazine, cis-1-Acetyl-4-[4-[[2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazine
CAS Number: 65277-42-1
MDL Number: MFCD00058579
Molecular Formula: C26H28Cl2N4O4
Molecular Weight: 531.43
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 150
Storage Temperature: 0-10°C
Product Categories: Bioscience, Antifungal Ingredients
Other Product Names / Synonyms: (+/-)-cis-1-Acetyl-4-[4-[[2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazine
Formula:
C₂₆H₂₈Cl₂N₄O₄ Melting Pt: 150 °C |
MDL Number:
MFCD00058579 CAS Number: 65277-42-1 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...