4-Isopropoxybenzenesulfonyl chloride ≥98.0%

Supplier: TCI America

Synonyms: 4-Isopropoxyphenylsulphonyl Chloride

I0802-25G I0802-5G
TCI0802-25GEA 234.3 USD
TCI0802-25G TCI0802-5G
4-Isopropoxybenzenesulfonyl chloride ≥98.0%
4-Isopropoxybenzenesulfonyl chloride

CAS Number: 98995-40-5
MDL Number: MFCD03093083
Molecular Formula: C9H11ClO3S
Molecular Weight: 234.69
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 113
Melting point (°C): 38
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 4-Isopropoxyphenylsulfonyl Chloride

Formula: C₉H₁₁ClO₃S
MW: 234.69 g/mol
Boiling Pt: 113 °C
Melting Pt: 38 °C
MDL Number: MFCD03093083
CAS Number: 98995-40-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b79f4c177fc881d25d9f457d61467dc9d6623c8f&vid=b79f4c177fc881d25d9f457d61467dc9d6623c8f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR