2-Iodofluorene ≥95.0% (by GC)
Supplier: TCI America
I0793-25G
I0793-5G
TCI0793-25GEA
247.9
USD
TCI0793-25G
TCI0793-5G
2-Iodofluorene ≥95.0% (by GC)
2-Iodofluorene
CAS Number: 2523-42-4
MDL Number: MFCD00019049
Molecular Formula: C13H9I
Molecular Weight: 292.12
Purity/Analysis Method:
Formula:
C₁₃H₉I Melting Pt: 130 °C |
MDL Number:
MFCD00019049 CAS Number: 2523-42-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...