4-Amino-1-isopropylpiperidine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
I0774-25G
I0774-5G
TCI0774-25GEA
326.4
USD
TCI0774-25G
TCI0774-5G
4-Amino-1-isopropylpiperidine ≥98.0% (by GC, titration analysis)
4-Amino-1-isopropylpiperidine
CAS Number: 127285-08-9
MDL Number: MFCD03411606
Molecular Formula: C8H18N2
Molecular Weight: 142.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 0.90
Formula:
C₈H₁₈N₂ MW: 142.2419 g/mol Density: 0.90 (20 °C) |
MDL Number:
MFCD03411606 CAS Number: 127285-08-9 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...