4-Isopropylpiperidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

I0733-1G I0733-5G
TCI0733-1GEA 137 USD
TCI0733-1G TCI0733-5G
4-Isopropylpiperidine ≥98.0% (by GC, titration analysis)
4-Isopropylpiperidine

CAS Number: 19678-58-1
MDL Number: MFCD09055342
Molecular Formula: C8H17N
Molecular Weight: 127.23
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 171
Specific Gravity (20/20): 0.86

Formula: C₈H₁₇N
Boiling Pt: 171 °C
Density: 0.86 (20 °C)
MDL Number: MFCD09055342
CAS Number: 19678-58-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5bdd96bc922af07d99262147364ddc23cd419908&vid=5bdd96bc922af07d99262147364ddc23cd419908&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR