3-Iodobenzoyl chloride ≥98.0%

Supplier: TCI America

Synonyms: m-Iodobenzoyl chloride

I0720-1G I0720-5G
TCI0720-1GEA 40 USD
TCI0720-1G TCI0720-5G
3-Iodobenzoyl chloride ≥98.0%
3-Iodobenzoyl chloride

CAS Number: 1711-10-0
MDL Number: MFCD02093934
Molecular Formula: C7H4ClIO
Molecular Weight: 266.46
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 146
Melting point (°C): 25

Formula: C₇H₄ClIO
MW: 266.47 g/mol
Boiling Pt: 146 °C
Melting Pt: 25 °C
MDL Number: MFCD02093934
CAS Number: 1711-10-0
UN: 3261
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90bb498f8d0f817c3653eecd63970b6d8971dda4&vid=90bb498f8d0f817c3653eecd63970b6d8971dda4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR