Isopentyl propionate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Isoamyl Propionate, Propionic acid isoamyl ester

I0677-25G I0677-500G
TCI0677-25GEA 21.37 USD
TCI0677-25G TCI0677-500G
Isopentyl propionate ≥98.0% (by GC)
Isopentyl propionate

CAS Number: 105-68-0
MDL Number: MFCD00048611
Molecular Formula: C8H16O2
Molecular Weight: 144.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 161
Flash Point (°C): 41
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: Propionic Acid Isoamyl Ester

Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 161 °C
Density: 0.87 (20 °C)
Flash Pt: 41 °C
MDL Number: MFCD00048611
CAS Number: 105-68-0
UN: 3272
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b10d3de321f1a08d2ca633aaedaeebaf97f013b5&vid=b10d3de321f1a08d2ca633aaedaeebaf97f013b5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR