3-Iodobenzonitrile ≥98.0% (by GC)
Supplier: TCI America
CAS Number: 69113-59-3
MDL Number: MFCD00079762
Molecular Formula: C7H4IN
Molecular Weight: 229.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 41
Flash Point (°C): 110
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 1-Cyano-3-iodobenzene
Formula:
C₇H₄IN Melting Pt: 41 °C Flash Pt: 110 °C |
MDL Number:
MFCD00079762 CAS Number: 69113-59-3 UN: 3439 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...