α-Bromo-3-iodotoulene ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 1-(Bromomethyl)-3-iodobenzene, α-Bromo-m-iodotoulene, 3-Iodobenzyl bromide

I0603-25G I0603-5G
TCI0603-25GEA 294 USD
TCI0603-25G TCI0603-5G
α-Bromo-3-iodotoulene ≥97.0% (by GC)
α-Bromo-3-iodotoulene

CAS Number: 49617-83-6
Molecular Formula: C7H6BrI
Molecular Weight: 296.93
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 52
Flash Point (°C): 113


Other Product Names / Synonyms: alpha-Bromo-3-iodotoluene

Formula: C₇H₆BrI
MW: 296.93 g/mol
Melting Pt: 52 °C
Flash Pt: 113 °C
MDL Number: MFCD00019016
CAS Number: 49617-83-6
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=951a4367f02e5085e4f771d4e46f55439fe0da42&vid=951a4367f02e5085e4f771d4e46f55439fe0da42&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR